Avagard™ Surgical and Healthcare Personnel Hand Antiseptic with Moisturizers, 3M™
Supplier: Healthcare Products
MMM 9201 MMM 9202 MMM 9216 MMM 9200
75787-528EA 153.67 USD
75787-528 75787-530 75830-242 75787-931
Avagard™ Surgical and Healthcare Personnel Hand Antiseptic with Moisturizers, 3M™
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
Avagard™ Surgical and healthcare personnel hand antiseptic combines alcohol for rapid kill, Chlorhexidine Gluconate for persistence plus emollients for skin conditioning.
  • Combines two active ingredients plus a lotion for effective antimicrobial activity and skin conditioning
  • Persistent antimicrobial protection
  • FDA approved as a health care personnel handwash
It is the only FDA NDA approved brushless, waterless, instant hand antiseptic that contains two active ingredients; alcohol for rapid kill and Chlorhexidine Gluconate for persistent, cumulativeMore Product Information