Biopsy Punch Tray, Sklar
Supplier: Sklar
96-1712
75818-008CS 794.6 USD
75818-008
Biopsy Punch Tray, Sklar
Trays/Dishes/Pots Instrument Trays/Dishes
Biopsy Punch Trays are industry standard
Each tray includes the following instruments and prep materials: 4mm Biopsy Punch, Webster Needle Holder, Curved Iris Scissors, Tissue Forceps, 30ga x 1/2" Needle, 25ga x 5/8" Needle, 3cc Syringe, Alcohol Prep Pad, PVP Ointment, (4) Gauze Sponges (2x2", 12-ply), Fenestrated Drape (18x26"), (2) Band-Aids, Gloves, and Tray.