Disodium hydrogen phosphate, anhydrous 98,0 FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic

S1402-12KG S1402-2.5KG S1402-45KGBL S1402-500GM
75812-290EA 1592.7 USD
75812-290 75812-292 75812-294 75812-296
Disodium hydrogen phosphate, anhydrous 98,0 FCC
Disodium hydrogen phosphate

Sodium Phosphate, Dibasic, Anhydrous, FCC is an additive used as an antioxidant, sequestrant and a pH regulator with very high buffering capacity widely used in foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
Merck Index: 12,08805

Order Now

Specification Test Results

Assay (Na₂HPO₄); Dried Basis) 98.0 % min.
Arsenic max.3 mg/kg
Fluoride max. 0.005 %
Insoluble Substances max. 0.2 %
Lead max. 4 mg/kg

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR