Trio.Bas™ DUO Kits
Supplier: Hardy Diagnostics
BAS220 BAS221 BAS226 BAS220K BAS225K BAS221K BAS226K
75804-716EA 9138.73 USD
75804-716 75804-718 75804-722 76076-708 76076-710 76169-646 76169-648
Trio.Bas™ DUO Kits
Samplers Air Samplers
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
  • Two aspirating heads with easy manipulation bayonet closure
  • Ability to use two different culture media at the same time
  • Save sampling time by doubling the aspirated volume of air
  • Manual or automatic operations
All of the air samplers are lightweight, with an antibacterial techno-polymer shockproof body. They have a long lasting battery and are IP65 protected from dust and water.
Per USP 797, 'Impaction is the preferred method of active air sampling. UseMore Product Information