VWR® Beakers with Handle, PP
Supplier: VWR International
75800-250 75800-252 75800-254 75800-256 75800-258 75800-260 75800-262
75800-250CS 58.03 USD
75800-250 75800-252 75800-254 75800-256 75800-258 75800-260 75800-262
VWR® Beakers with Handle, PP
Beakers
VWR® polypropylene beakers, which come in a natural color, have a handle, pouring spout and graduations printed directly on the beakers.
  • Autoclavable
  • Integrated pouring lip
  • Natural color
  • Printed graduation
The beakers are autoclavable. Working temperatures of 0 to 121 °C.
Certificaciones: Resin meets US FDA 21 CFR requirements.
Precaución: Do not stack products while autoclaving.