Water, Cell Culture grade

Supplier: GROWCELLS

Synonyms: Hydrogen oxide

CCPW-2000 CCPW-010L CCPW-1000 CCPW-0125 CCPW-0500 CCPW-020Q
10218-876CS 502.25 USD
10218-876 10218-880 10218-882 10220-414 75788-536 75788-538 75788-540 75792-294 75792-296
Water, Cell Culture grade
Water

Distilled cell culture grade water.


  • Prepared by distillation
  • Steam sterilization in final packaging
  • 0.1 micron sterile filtration


Distilled cell culture grade water, <0.005EU/mL. Prepared by distillation, 0.1 micron sterile filtration and steam sterilization in final packaging.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR