Pro-Loops Nichrome Wire Loops, Pro-Lab Diagnostics
Supplier: PRO-LAB DIAGNOSTICS INC CA
PL.120 PL.100 PL.101 PL.102 PL.103 PL.104 PL.105 PL.130 PL.131 PL.133
75784-796PK 330.41 USD
75784-796 75784-808 75784-810 75784-812 75784-814 75784-820 75784-824 75784-870 75784-872 75784-874
Pro-Loops Nichrome Wire Loops, Pro-Lab Diagnostics
Spreaders Inoculating Loops and Needles
When strict calibration is required, certified/calibrated loops from Pro-Lab Diagnostics are manufactured from the highest quality 80/20 nickel chromium wire.
  • Useful for strict calibration
  • Manufactured from the highest quality, 80/20 nickely chromium wire
  • Presented in a protective case
When strict calibration is required, certified/calibrated loops from Pro-Lab Diagnostics are manufactured from the highest quality 80/20 nickel chromium wire and calibrated using the Evans Blue Dye Method. Each pack of five loops is presented in a protective case andMore Product Information