Single Taper Inlet Liner with Wool for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets, Restek
Supplier: Avantor
75778-780PK 290.12 USD
75778-780
Single Taper Inlet Liner with Wool for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.