SP Bel-Art Biohazard Disposal Can with Lift-up Cover, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13197-0000
74950-062EA 82.98 USD
74950-062
SP Bel-Art Biohazard Disposal Can with Lift-up Cover, Bel-Art Products, a part of SP
Bins Waste Bins
Bright red safety disposal container is suitable for biological/biohazard materials, broken glass, recyclables or other waste and sized for benchtop or floor placement.
  • Polypropylene construction
  • Long service life
  • Puncture-resistant
  • Autoclavable
  • One-piece hinged lid contains odors and protects users from contents
Molded of polypropylene for long service life, this can is puncture-resistant and autoclavable at 121°C (250°F). One-piece hinged lid protects users from contact with the contents, helps retain odors and contains airborne contaminants.