Nalgene™ Safety Waste Funnels, Thermo Scientific
Supplier: Thermo Fisher Scientific
6378-0004 6378-0010
73520-198CS 721.78 USD
73520-198 73520-200
Nalgene™ Safety Waste Funnels, Thermo Scientific
Funnels Safety Funnels
HDPE funnels with attached lid, polypropylene closure/funnel adapter, screen insert, polypropylene/PTFE 0.45µm vent plug, and filter. For use with Thermo Scientific™ Nalgene™ Safety Waste Funnel Systems.
  • Funnels feature the Easy Snap Safety Latch to minimize spillage if accidentally tipped over and a larger diameter funnel for easy pouring
  • The vent system helps reduce fumes and splashing while the removable neck screen traps large particles, like stir bars
  • Designed for solvent, chemical and biological waste, it is recommended for use with Nalgene
More Product Information