Nalgene® Buckets, White Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
7012-0110 7012-0080 7012-0140
17306-005CS 418.58 USD
17306-005 73470-016 73470-018
Nalgene® Buckets, White Polypropylene, Thermo Scientific
Buckets
Crack-resistant polypropylene buckets can be autoclaved or gas sterilized for use in the food industry
Note: Do not sterilize with radiation or dry heat. Not suitable for strong alkalis.
Durable enough for handling photographic chemicals and most corrosive liquids. Nylon-coated wire bail handle. Autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable