HLT Titertek 200 Specialty Tips, Molecular BioProducts
Supplier: Thermo Scientific
3850-11
72830-676CS 764.67 USD
72830-676
HLT Titertek 200 Specialty Tips, Molecular BioProducts
Pipette Tips
Titertek style pipettors 20-200 ul, conatins 1000 tips in each bag.
  • Clear
  • Non-sterile
  • Nucleic acid and nuclease free
  • Non pyrogenic