Ethidium bromide

Supplier: G-Biosciences

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

RC-049
71003-400EA 201.31 USD
71003-400
Ethidium bromide
Ethidium bromide

G-Biosciences' Ethidium Bromide is supplied either as a substance or as a mixture (solution). Two concentrations of ethidium bromide solutions are offered: a 0.625 mg/mL solution and a 10 mg/mL solution.

Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Boiling Pt: 72
MDL Number: MFCD00011724
CAS Number: 1239-45-8
Merck Index: 13,04751

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR