Orthoboric acid for biochemistry

Supplier: G-Biosciences
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

RC-029 RC-028
71003-368EA 111.86 USD
71003-368 71003-366
Orthoboric acid for biochemistry
Orthoboric acid
Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185
Melting Pt: 160
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR