Trypsinizing Flasks, Baffled, with Sidearm and Screw Caps , Ace Glass Incorporated
Supplier: Ace Glass
3902-34 3902-36 3902-32
71000-494CS 499.94 USD
71000-494 71000-500 71000-508
Trypsinizing Flasks, Baffled, with Sidearm and Screw Caps , Ace Glass Incorporated
Flasks Trypsinization Flasks
Trypsinizing flask for the converting of homogenous tissue samples into cell suspensions by digestion of connective tissue
Side port has dam below the sidearm to prevent undigested tissue from being poured out while decanting cells. Flask has GPI thread top and side arm cap. The three extra deep, extra long side baffles greatly enhance vigorous agitation.
GPI Thread Finish: 38-430
Sidearm Thread: 20-415