Orange II

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Tropaeolin OOO-2, acid orange 7, acid orange A, D and C Orange No. 4, 4-(2-Hydroxy-1-naphthylazo)benzenesulphonic acid sodium salt, Orange II sodium salt, p-(2-Hydroxy-1-naphthylazo)benzenesulphonic acid sodium salt, Gold orange

D1130-500GM D1130-100GM
700010-014EA 1546.7 USD
700010-014 700010-010
Orange II
Orange II

D and C Orange No. 4 | Non-graded D and C Orange No. 4 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


GHS hazard : Exclamation mark

Formula: C₁₆H₁₁N₂NaO₄S
MW: 350.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011657
CAS Number: 633-96-5

Order Now

Specification Test Results

Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR