Iron(III) citrate monohydrate, purified

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Iron (III) citrate tribasic monohydrate, Ferric citrate monohydrate, Ferric Citrate, n-Hydrate

F1028-125GM F1028-12KG F1028-500GM F1028-2.5KG F1028-50KGBL
700006-190EA 157.61 USD
700006-190 700006-192 700006-196 700006-194 700006-198
Iron(III) citrate monohydrate, purified
Iron(III) citrate monohydrate

Ferric Citrate, Purified is a powder with a green to reddish-brown appearance. It can also be used in water purification, printing and an acidity regulator.

Formula: C₆H₅FeO₇·H₂O
MW: 262.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013098
CAS Number: 2338-05-8
EINECS: 249-117-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR