Trichloroacetic acid (TCA) 50% w/v aqueous solution

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: TCA

T-182-500MLG
700004-674EA 421.33 USD
700004-674
Trichloroacetic acid (TCA) 50% w/v aqueous solution
Trichloroacetic acid (TCA)

Trichloroacetic Acid, 50 Percent (w/v) Aqueous Solution also known as TCA, is used in biochemistry to help the precipitation of macromolecules and in chemical peels to treat fine wrinkles, brown spots, mild acne, blotchy skin and for wart removal. Superficial TCA peels are up to 20 percent strength and can be performed every few months to achieve a more youthful appearance. Peels at 20 - 50 percent strength should be performed only in a doctor's office and are often done under general anesthesia.

Formula: Cl₃CCOOH
MW: 163.39 g/mol
Boiling Pt: 196
Melting Pt: 54…56 °C
Density: ≤1.07 g/cm³ (20 °C)
Flash Pt: 113
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 2564
ADR: 8 II
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR