Fish oil, from menhaden

Supplier: SPECTRUM CHEMICAL MFG CORP

F1192-500ML
700000-100EA 93.7 USD
700000-100
Fish oil, from menhaden
Fish oil, from menhaden

Fish Oil, From Menhaden is an oil obtained from the small North Atlantic fish, Brevoortia tyrannus. Fish oil derived from Menhaden boast low levels of mercury while still being high in omega-3 fatty acid. It is used in cosmetic and personal care products as a skin-conditioning agent, emollient and solvent. It appears as a liquid that is insoluble in cold and hot water

Density: 0.930
Storage Temperature: Freezer
MDL Number: MFCD00131085
CAS Number: 8002-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR