SlimLine™ Stainless Steel Cart Systems, Micronova
Supplier: Micronova
C-22S C-33S C3A-03 C-227
68400-726EA 3105.11 USD
68400-726 68400-728 94023-746 89423-426
SlimLine™ Stainless Steel Cart Systems, Micronova
Mops
Durable cart systems feature a stainless steel frame with push bar and three grip-it holders for easy storage and interchange of handles and additional mop heads. Systems come standard with color-coded, 22.7 liter (6 gal.) polypropylene buckets. \
  • Crafted from 304 electropolished stainless steel for durability and long life
  • Angled push bar for easy handling in the cleanroom
  • Color-coded, 22 L (6 gallon) polypropylene, or 30 L (8 gallon) stainlesss steel buckets included
  • Optional wringer bar allows wringers to be placed above the rinse bucket
  • Prevenz™
More Product Information