CGT™ Conductive Grid Tapes, Micronova
Supplier: Micronova
CGT-38 CGT-12 CGT-1 CGT-15
68399-986CS 947.78 USD
68399-986 68399-988 68399-992 68399-994 68399-998
CGT™ Conductive Grid Tapes, Micronova
Tapes
Conductive, anti-static polypropylene tape, specially designed with conductive grids embedded between two anti-static layers, for use on devices in ESD sensitive environments.
  • Heavy-duty, anti-static copolymer tape with acrylic adhesive
  • Manufactured for use in ESD sensitive environments
  • Features a conductive grid embedded between two antistatic layers
  • Conducts charge away from substrate
  • Protects sensitive devices that would be otherwise damaged by normal insulating tape
  • Provides an effective static shield and prevents static buildup
  • Prevents shedding,
More Product Information