Amplitude™ ProZorb® Wipers, Contec®
Supplier: Contec
PZ-44 PZ-99 PZ-1212
68310-206CS 396.78 USD
68310-206 68310-208 68310-210
Amplitude™ ProZorb® Wipers, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Amplitude™ Prozorb® wipes are a hydroentangled blend of cellulose and polyester, produced through a special process that creates apertures in the fabric while enhancing the thickness and providing excellent cleanliness.
  • Recommended for ISO Class 5 - 6 environments
  • Textured surface promotes effective cleaning and particle removal
  • Excellent sorbency, greatest level of intrinsic sorbency, and quickest rate of sorbency of any similar weight nonwoven wipe
  • Low levels of particles and extractables
  • Great for spill cleanup and wiping to dry in critical environments
More Product Information