Dextrose solution 5% USP for injection

Supplier: BAXTER IV SOLUTIONS

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

2B0062Q
68000-132CS 715.89 USD
68000-132
Dextrose solution 5% USP for injection
D-(+)-Glucose

  • Total calories: 170 kcal/l
  • Specific gravity: 1.02
  • Fill range volume (ml): 265 to 285
  • Store at room temperature (25 °C)
  • pH : 4.0 (3.2 - 6.5)
  • mOsmol/L (calc): 252
  • NDC Number: 0338-0017-02


Certificaciones: USP specified.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
MDL Number: MFCD00148912
CAS Number: 50-99-7
Merck Index: 14,04459

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR