Sugar Refractometer, 0 to 32%, Sper Scientific
Supplier: Sper Scientific
300001 300010
66214-978EA 213.46 USD
66214-978 66214-988
Sugar Refractometer, 0 to 32%, Sper Scientific
Refractometers Handheld Refractometers
Brix refractometers, also called sugar refractometers, are commonly used in the food and beverage, agriculture, and wine industries
A refractometer measures the extent to which light is bent (refracted) through a liquid sample. They are commonly used to help identify or confirm the identity of a sample, assess the purity of a sample, or determine the concentration of a solute in a solution.
It is the latter application for which this Sper Scientific handheld sugar-brix refractometer is an ideal measurement instrument for the food and beverage industry. Simply place a drop or two ofMore Product Information