Coliwasa Trumpet, Disposable, Wheaton®
Supplier: DWK Life Sciences (KIMBLE)
885235
66174-235CS 417.92 USD
66174-235
Coliwasa Trumpet, Disposable, Wheaton®
Samplers Disposable Samplers
Use for sampling sludge or semi-solid material.
Disposable glass coliwasa will bore through semi-solid sludge type samples. Liquid and sludge material will remain in the coliwasa due to its tight PTFE resin seal. Tube is 101.6 cm long with 200 ml capacity.
Order Now
SPECIFICATIONS
Environmentally Preferable