Pliers, Terminal Plug Forming, Excelta
Supplier: Excelta
907-101
63043-020EA 613.08 USD
63043-020
Pliers, Terminal Plug Forming, Excelta
Pliers
Designed to insert or extract the hard to reach terminal plugs.
  • Forms a terminal plug
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
The 907-101 five star 6" terminal handling plier is designed to insert or extract the hard to reach terminal plugs. The inside jaw has a cutout to prevent nicking or pulling on the lead wire. It features soft cushioned ergonomic grips (10¹⁰ ohms/sq surface resistivity) with dual leaf springs for smooth action.
Order Now
SPECIFICATIONS
Environmentally Preferable