Wire Stripper, Excelta
Supplier: Excelta
502E-US 510E-US
63042-432EA 427.45 USD
63042-432 63042-500
Wire Stripper, Excelta
Pliers
These two tools are designed for stripping PTFE coated small wires.
  • Will strip PTFE coating in one shot
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These tools are made to strip small PTFE coated wire up to 30AWG. The 502-US will strip one size wire; (specify the wire gauge size by indicating the gauge at the end of the reference number. Available in 22, 24, 26, 28, and 30 AWG.) The 510E-US is a 'V' notched wire stripper which cuts the insulation on four sizes makeing stripping much easier. It features a ergonomically moldedMore Product Information