Anti-TPX2 Mouse Monoclonal Antibody [clone: 18D5]

Supplier: Biolegend
628002
628002-BLEA 257.8 USD
628002-BL
Anti-TPX2 Mouse Monoclonal Antibody [clone: 18D5]
Antibodies
Purified anti-TPX2 [18D5]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IHC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TPX2
Clonality: Monoclonal
Clone: 18D5
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR