Falcon® CultureSlides, Corning
Supplier: Corning
354118 354114 354104 354108
53106-306CS 346.14 USD
53106-306 53106-304 62405-176 62405-178
Falcon® CultureSlides, Corning
Slides Chamber Slides
Soda-lime glass slide, PS vessel, lid and tool for loosening chamber.
  • Chamber easily and safely removed using the disposable safety removal tool
  • Pressure-sensitive, biocompatible, acrylic-adhesive gasket remains with the vessel after removal, not on the slide, facilitating further processing or placement of coverslips
  • Blue hydrophobic border defines cell culture areas
  • Wells numbered for easy identification
  • Trays designed for incubator use
Falcon® CultureSlides allow cells to be cultured and then analyzed on a glass microscope slide. Cells are grown inMore Product Information