Magna Flex™ Microcarrier Spinner Flask, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
356830 356831 356832 356834 356837 356839
62402-240CS 392.79 USD
62402-240 62402-242 62402-244 62402-246 62402-248 62402-250
Magna Flex™ Microcarrier Spinner Flask, Wheaton
Flasks Spinner Flasks
Designed for microcarrier cultures with a magnetic stirrer
  • Gentle stirring action provided by a bulb shaped glass impeller
  • Flasks provides head space ratio of 1:1 for ideal gas exchange
Made from Autoclavable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable