PCR-Cooler and PCR Racks
Supplier: Eppendorf
022510541 022510509 022510525 0030124545
62111-758EA 112.96 USD
62111-758 62111-762 62111-760 89135-368
PCR-Cooler and PCR Racks
Coolers Benchtop Coolers
Eppendorf PCR-Racks for safe PCR sample storage/transport with color coding for easy identification - ideal for 0.1, 0.2, 0.5 ml tubes/strips. The PCR-Cooler keeps samples cool for 1hr+ post 2hrs in -20 °C, with temperature indicator for sample safety.
  • Handling system for sample set-up, protection, transport, and storage of sensitive samples - keeps your samples safe
  • Clear temperature indicator: Color of PCR-Cooler changes when temperature exceeds 7 °C
  • Accommodates PCR vessels as tubes, strips, or plates for flexible vessel usage
  • Dry incubation technology
More Product Information