Barnstead/Thermolyne Temperature Controller, Thermo Scientific
Supplier: Thermo Fisher Scientific
CN45515
61329-010EA 330.38 USD
61329-010
Barnstead/Thermolyne Temperature Controller, Thermo Scientific
Controllers Temperature Controllers
Controls power input to heating tapes and heating mantles.
  • Utilizes time-on/time-off stepless input control
  • Compensates for line voltage fluctuations and ambient temperature changes
  • Built-in cycle and power-on lights
  • Single duplex outlet at rear of housing
Maximum load: 15 A.
Información de suministro: Complete with 15.2 m (50') three-wire cord and plug.