VWR® Traceable® Solar-Powered Thermometers
Supplier: VWR International
61161-334
61161-334EA 77.3 USD
61161-334
VWR® Traceable® Solar-Powered Thermometers
Thermometers Digital Thermometers
Thermometers are “always on” and run for years powered from light in the lab.
  • Traceable®
  • Solar-powered
  • Waterproof
  • Reads in °C and °F
Choose thermometer with internal sensor to monitor ambient temperature or thermometer with external 0.5 cm (3/16") diameter sensor and 304.8 cm (10') long cable to monitor temperatures of liquids, air/gas, and semi-solids
Units sit on the bench and incorporate a slot for wall mounting. Read °F or °C at the flip of a switch. The 1.3 cm (1/2") LCD updates every 10 seconds and is readable from 10 feet.