SP Bel-Art Thermometer Rack, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18981-0001
61136-550CS 546.81 USD
61136-550
SP Bel-Art Thermometer Rack, Polypropylene, Bel-Art Products, a part of SP
Thermometer Holders
This polypropylene rack holds up to 25 thermometers in correct vertical position to prevent separation which may occur when thermometers are stored improperly.
  • Polypropylene construction
  • Hold thermometers vertically to prevent separation
  • Autoclavable
Hole diameters in the top and middle plates are 7.9 mm (516") while the bottom plate has 3.2 mm (1/8") diameter drain holes. The distance between top and bottom plates is 10.5 cm (41/8"). Steam autoclavable at 121 °C (250 °F).
Order Now
SPECIFICATIONS
Environmentally Preferable