VWR® Plate Paddles
Supplier: VWR International
60941-128
60941-128CS 128.39 USD
60941-128
VWR® Plate Paddles
Film Applicators Sealing Film Applicators
An easy alternative to rollers, these plate paddles are designed to provide secure sealing around all wells on a microplate.
  • Alternative to rollers
  • Secure seal around all microplate wells
  • Especially effective with raised-rim microplates
Paddles are especially recommended when applying films to raised-rim microplates due to their precise fit within the rim of the plate.
Order Now
SPECIFICATIONS
Environmentally Preferable