Histosette® II Tissue Processing/Embedding Cassettes with Integrated Lids, Simport
Supplier: Simport Scientific
M492-16 M492-3 M492-5 M492-6 M492-7 M492-8 M492-9 M492-10 M492-11 M492-2 M492-4 M492-12 M492-18
87002-454CS 456.15 USD
87002-454 87002-456 60872-520 60872-524 60872-526 60872-528 60872-530 60872-532 60872-534 60872-536 60872-518 60872-522 87002-452 10040-366
Histosette® II Tissue Processing/Embedding Cassettes with Integrated Lids, Simport
Embedding Cassettes Tissue Processing Cassettes
These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage
Molded from a special high-density polymer, they can keep specimens safely submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lids can be opened and closed as often as necessary and will always relock securely without danger of specimen loss. The unique front hinge design allows for single-hand cassette manipulation. The anterior writingMore Product Information