SGE Syringes, General Purpose Manual Syringe, PTFE Tipped Plunger, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
002202 002208 002252 002313 002315 002500 003312 004312 005312 006312 007312 008502 008962 009463 002330 002335 008140 002200 002250 002301 003200 004200 004230 004250 005200 005230 005250 006200 006230 006250 007200 007230 008025 008425 008760 008960 009462 009660 009760 004229 005229 006229 007229 008020 008420 008100 008500 008700 008900 002455 008505 002300 008105 008102 003250 007250
14225-198PK 437.56 USD
14225-198 14225-200 14225-202 14225-204 14225-206 14225-216 14225-248 14225-282 14225-312 14225-338 14225-364 14225-398 14225-412 14225-422 14227-512 14227-514 14233-486 60361-092 60361-094 60361-096 60361-146 60361-160 60361-162 60361-164 60361-170 60361-172 60361-174 60361-178 60361-180 60361-182 60361-188 60361-190 60361-196 60361-200 60361-204 60361-208 60361-212 60361-216 60361-220 60361-432 60361-434 60361-436 60361-438 60361-440 60361-442 60361-444 60361-446 60361-448 60361-450 60361-452 60361-472 60361-474 60361-476 14225-378 60361-148 60361-192
SGE Syringes, General Purpose Manual Syringe, PTFE Tipped Plunger, Trajan Scientific and Medical
Syringes Syringes for Chromatography
SGE general purpose manual syringes feature Diamond Syringe Technology.
PTFE Tipped Plunger Syringes, commonly known as gas tight syringes, are available from 10µL to 100mL and are the ideal choice for dirty samples, liquid or gas samples
Enhancements to SGE Diamond syringes result in improved solvent resistance, greater temperature ranges and increased operational smoothness. SGE Diamond manual syringes offer a new level of accuracy due to reduced carryover and eliminates the risk of adhesive contamination. The syringes are available in:al purpose manual syringes feature DiamondMore Product Information