Synthware Friedrichs Condensers with Removable Hose Connections, Kemtech America
Supplier: Kemtech America
C272440
60000-822EA 235.22 USD
60000-822
Synthware Friedrichs Condensers with Removable Hose Connections, Kemtech America
Condensers
With a joint at a 15° angle.
  • Nylon caps and hose connection included
  • Friedrichs style condenser
  • Heavy wall borosilicate construction
  • Ground glass joint
Supplied complete with nylon cap and hose connections. Hose connection size: 8 mm.