VWR® Spinbar® Stir Bars, Cylindrical
Supplier: VWR International
58947-150 58948-047 58948-058 58948-952 58948-954 58948-956 58948-958 58948-960 58948-962 58948-964 58948-966 58948-968 58948-970 58949-061 58949-083 58949-094 58949-108 58949-130 58949-152
58947-150CS 643.43 USD
58947-150 58948-047 58948-058 58948-952 58948-954 58948-956 58948-958 58948-960 58948-962 58948-964 58948-966 58948-968 58948-970 58949-061 58949-083 58949-094 58949-108 58949-130 58949-152
VWR® Spinbar® Stir Bars, Cylindrical
Stirrers Magnetic Stirring Bars
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics.
  • Small removable pivot ring in the center adds to their versatility
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating
  • Durable
  • Inert
  • Manufactured in a registered ISO 9001:2008, as verified by SGS certification
  • Autoclavable
Pivot ring minimizes the contact area of the bar to the vessel, reduces friction and lessens marring of plastic containers (7.9x12mm and 7.9x19mm models have no pivot ring).
Información para pedidos:More Product Information