VWR® Spinbar® Magnetic Stir Bars, Octagon
Supplier: VWR International
66001-158 80089-636 58947-106 58947-114 58947-120 58947-122 58947-124 58947-128 58947-132 58947-136 58947-138 58948-025 58948-070 58948-091 58948-116 58948-138 58948-150 58948-171 58948-193 58948-218 58948-230 58948-251 58948-273 58948-309 58948-950
66001-158PK 443.01 USD
66001-158 80089-636 58947-106 58947-114 58947-120 58947-122 58947-124 58947-128 58947-132 58947-136 58947-138 58948-025 58948-070 58948-091 58948-116 58948-138 58948-150 58948-171 58948-193 58948-218 58948-230 58948-251 58948-273 58948-309 58948-950
VWR® Spinbar® Magnetic Stir Bars, Octagon
Stirrers Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
  • Pivot ring aids in reducing friction and chattering (58948-091 does not have a pivot ring)
  • Autoclavable
  • Manufactured in a registered ISO 9001:2008, as verified by SGS certification
Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars.
Información para pedidos: 6-Assortment includesMore Product Information