Zero Charge® Hand Lotion, Tech Spray
Supplier: ITW Chemtronics
1702-8FP
58542-419CS 20.45 USD
58542-419
Zero Charge® Hand Lotion, Tech Spray
Skincare Lotions Hand and Body Lotions
This lotion contains a special blend of moisturizers (including aloe vera and vitamins A and D) that leave skin soft and nongreasy
It is also an antistatic formulation designed to improve contact between the user’s skin and grounding devices, such as wrist and heel straps, to prevent static build-up. Absorbs completely. Does not contain silicone, lanolin, squalene, or similar materials found in conventional hand lotions that can interfere with PCB manufacturing.