VWR® Test Sieves 8", Stainless Steel
Supplier: VWR International
57334-385 57334-386 57334-387 57334-388 57334-392 57334-394 57334-396 57334-398 57334-400 57334-404 57334-406 57334-408 57334-410 57334-412 57334-414 57334-416 57334-418 57334-420 57334-422 57334-424 57334-426 57334-430 57334-432 57334-434 57334-436 57334-438 57334-440 57334-442 57334-444 57334-446 57334-448 57334-450 57334-452 57334-454 57334-456 57334-458 57334-460 57334-462 57334-464 57334-466 57334-468 57334-470 57334-472 57334-474 57334-476 57334-478 57334-480 57334-482 57334-484 57334-486 57334-488 57334-490 57334-492 57334-494 57334-504 57334-516 57334-518 57334-520 57334-522 57334-526 57334-528 57334-530 57334-532 57334-534 57334-536 57334-538 57334-540 57334-542 57334-544 57334-546 57334-548 57334-550 57334-552 57334-554 57334-556 57334-558 57334-560 57334-562 57334-566 57334-568 57334-570 57334-572 57334-574 57334-576 57334-580 57334-582 57334-584 57334-586 57334-588 57334-590 57334-592 57334-594 57334-596 57334-598 57334-600 57334-602 57334-604 57334-402 57334-428 57334-564 57334-578
57334-385EA 340.13 USD
57334-385 57334-386 57334-387 57334-388 57334-392 57334-394 57334-396 57334-398 57334-400 57334-404 57334-406 57334-408 57334-410 57334-412 57334-414 57334-416 57334-418 57334-420 57334-422 57334-424 57334-426 57334-430 57334-432 57334-434 57334-436 57334-438 57334-440 57334-442 57334-444 57334-446 57334-448 57334-450 57334-452 57334-454 57334-456 57334-458 57334-460 57334-462 57334-464 57334-466 57334-468 57334-470 57334-472 57334-474 57334-476 57334-478 57334-480 57334-482 57334-484 57334-486 57334-488 57334-490 57334-492 57334-494 57334-504 57334-516 57334-518 57334-520 57334-522 57334-526 57334-528 57334-530 57334-532 57334-534 57334-536 57334-538 57334-540 57334-542 57334-544 57334-546 57334-548 57334-550 57334-552 57334-554 57334-556 57334-558 57334-560 57334-562 57334-566 57334-568 57334-570 57334-572 57334-574 57334-576 57334-580 57334-582 57334-584 57334-586 57334-588 57334-590 57334-592 57334-594 57334-596 57334-598 57334-600 57334-602 57334-604 57334-402 57334-428 57334-564 57334-578
VWR® Test Sieves 8", Stainless Steel
Sieves Test Sieves
VWR® 8" diameter (203.2 mm) all stainless steel test sieves are durable and ideal for sieving applications where a medium sample size is required.
  • Highly durable construction
  • Stainless steel fabrication
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are all available accessories.
Información accesorios: Half height (More Product Information