SP Bel-Art® Safety Acid/Caustic Samplers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37801-0000 H37802-0000
56615-730EA 453.02 USD
56615-730 56615-731
SP Bel-Art® Safety Acid/Caustic Samplers, Bel-Art Products, a part of SP
Samplers Fluid Samplers
Excellent for sampling from tank cars, tank trucks and barges, these samplers are well suited for sodium hydroxide, potassium hydroxide, hydrochloric acid, acetic acid, formaldehyde, sulfuric acid, etc.
  • Polyethylene plastic construction
  • Slot for fluid entry into bottle
  • Upper end of the bar is threaded to accept the handle
  • Ideal for sampling caustic liquids
A 500cc (16oz) polyethylene bottle is screwed onto a 10 cm (4”) long, 38 mm (1½”) diameter bar which has a slot for fluid entry
500cc (16 oz.) replacement bottles: catalog no. 47750-612.