Drum Storage Cabinets, Eagle Manufacturing
Supplier: Eagle Manufacturing
1928X 1926X 2610X 2810X
56610-618EA 3607.74 USD
56610-618 56610-629 56610-637 56618-566
Drum Storage Cabinets, Eagle Manufacturing
Furniture Storage Cabinets
Perfect for safe storage of flammable liquid drums near work areas.
  • Finish is caution-yellow enamel with red warning
  • Complete with grounding attachments, three-point key lock, and 5 cm (2") raised leakproof door sill
  • Drum cabinet ramp attaches firmly to cabinet sill and accommodates most dollies for simplified loading and unloading
  • Ramp is constructed of heavy duty galvanized steel, and can withstand loads up to 680 kg (1500 lbs.)
Horizontal storage cabinet interior is equipped with drum cradle guide rails. Rails are 13.9 cm (5½") wide and are set 27.9More Product Information