VWR® CryoPro® Canister Storage Tanks, CC Series

Supplier: VWR International
CC-3 CC-4 CC-2 CC-5
55709-216EA 3118.51 USD
55709-216 55709-220 55709-222 55709-218 55709-224
VWR® CryoPro® Canister Storage Tanks, CC Series
Cryopreservation Systems
For small to medium capacity applications, the CC series offers traditional canister-style cryogenic storage in efficient narrow neck designs.

  • Sealed shaft prevents hazardous spurting of LN2
  • Aluminum canes simplify and organize cryogenic vials for storage

CryoRetriever claw-style tool (82017-944) is specifically designed for retrieving boxes, vials, and frames from cryopreservation tanks

Sturdy five-wheel rollerbases are available for safely moving full tanks. A low liquid level alarm (55709-276) helps protect against accidental warmups. The phaseMore Product Information
Order Now

SPECIFICATIONS

Model CC-3 CC-4 CC-2 CC-5
Environmentally Preferable
Canister Height 27.9 cm (11") 27.9 cm (11") 27.9 cm (11") 27.9 cm (11")
Canister ∅ 5.6 cm (21/4") 7.1 cm (213/16") 4.2 cm (15/8") 10.2 cm (4")
Evaporation Rate 0.14 L/Day 0.18 L/Day 0.09 L/Day 0.39 L/Day
Ext.Ø 46.2 cm (183/16") 46.2 cm (183/16") 36.8 cm (141/2") 50.8 cm (20")
Height, with Lid 66 cm (26") 67.5 cm (269/16") 65.2 cm (2511/16") 67.3 cm (261/2")
Liquid Nitrogen Capacity 33.4 L 34.8 L 20.5 L 47.4 L
Neck Int.Ø 7 cm (23/4") 8.9 cm (31/2") 5.5 cm (23/16") 12.7 cm (5")
Weight Empty 15.4 kg (34 lbs.) 15.4 kg (34 lbs.) 11.8 kg (26 lbs.) 16.3 kg (36 lbs.)
Weight Full 42.5 kg (93.7 lbs.) 43.5 kg (95.9 lbs.) 28.3 kg (62.4 lbs.) 54.6 kg (120.4 lbs.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR