NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
4020.X00V0 4050.X00V0 4200.X00V0 NJ-4606701-02 NJ-4606710-02 NJ-4606728-02 NJ-4606736-02
80076-428CS 536.88 USD
80076-428 53548-017 53548-019 53548-025 80076-426 76459-484 76459-486 76459-488 76459-490
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Syringes Syringes, Medical
NORM-JECT® syringes feature medical-grade plastics, including a polypropylene barrel and a polyethylene plunger. Great value at reasonable costs.
  • Sterile
  • Certified DNA free
  • Latex free
  • Rubber free
  • Silicone free
  • Styrene free
  • DEHP free
These syringes are the choice for any situation needing an inert, non-reactive syringe. Because of their composition, these syringes are indicated for Nuclear Medicine, Amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures. They are more chemically resistant than rubber tipped syringes. TheseMore Product Information