Upchurch Scientific® Stainless Steel Unions, IDEX Health & Science
Supplier: Upchurch Scientific
U-322 U-402 U-437 U-412 U-435 UH-402
21511-306EA 77.08 USD
21511-306 21511-316 53500-820 21511-322 21511-330 97055-126
Upchurch Scientific® Stainless Steel Unions, IDEX Health & Science
Fittings and Connectors
Upchurch Scientific® high-pressure, zero dead volume unions manufactured by IDEX Health & Science, are precision machined from 316 stainless steel, carefully passivated, and thoroughly rinsed.
  • High-pressure rating
  • Zero dead volume
  • Precision machined from 316 stainless steel
Each Union comes complete with stainless steel nuts and ferrules for 1/16 in. or 1/8 in. OD tubing, and all are pressure rated to 20,000 psi (1,380 bar) or higher. In addition to our standard unions, we also offer several special versions: a VICI® (Valco) compatible version; aMore Product Information