Autoclave Bag Red with Heat Indicator 300°F, Hardy Diagnostics

Supplier: Hardy Diagnostics
AB5013
52428-866PK 175.91 USD
52428-866
Autoclave Bag Red with Heat Indicator 300°F, Hardy Diagnostics
Bags Sterilization Pouches
Autoclave Bag is made of tough, thick polypropylene, double seamed bottom, solid red bag with black biohazard warnings in English and Spanish.

  • Thickness 2 mil
  • With heat indicator
  • Bag is rated to withstand heat up to 300 °F

Storage temperature: 15...30 °C. Heat indicator strip turns black when exposed to steam, OSHA approved labeling, plastic twist ties for closure furnished upon request.

Envase: 200 per case.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR