Anti-IL-17A Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: TC11-18H10.1]

Supplier: Biolegend
506929
506929-BLEA 267 USD
506929-BL
Anti-IL-17A Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: TC11-18H10.1]
Antibodies
Brilliant Violet 650™ anti-mouse IL-17A [TC11-18H10.1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 125 μl

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-17A
Clonality: Monoclonal
Clone: TC11-18H10.1
Conjugation: Brilliant Violet® 650
Epitope:
Host: Rat
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR