Anti-IFN-g Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: B27]
Supplier: Biolegend
506517 506518
506517-BLEA 157.7 USD
506517-BL 506518-BL
Anti-IFN-g Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: B27]
Antibodies
PE/Cy7 anti-human IFN-γ [B27]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, African Green Monkey, Pigtailed Macaque, Sooty Mangabey; Apps: ICFC; Size: 100 tests
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagentMore Product Information